Learn More
Electron Microscopy Sciences Cryo Gel, 4.3oz/Dispenser 4.3 OZ
SDP
Catalog No. 5029319
Encompass
Crogenic gel is a highly viscous, water-soluble embedding medium for frozen sections that "stays put". The gel is packaged in a pump-dispenser bottle that can instantly dispense the gel in any position. The dispenser has a built-in "guillotine" that cleanly severs the gel from the pump spouts and prevents any "stringy" medium from pulling away when the bottle is removed.
To use, simply dispense gel on a room temperature block-holder. Use a cold heat extractor to form a base layer. Dispense additional gel. Place the tissue on top of the gel. Use a cold heat extractor to freeze tissue and form a block.
Catalog No. 50-293-19 Supplier Electron Microscopy Sciences Supplier No. 6280601
Crogenic gel is a highly viscous, water-soluble embedding medium for frozen sections that "stays put". The gel is packaged in a pump-dispenser bottle that can instantly dispense the gel in any position. The dispenser has a built-in "guillotine" that cleanly severs the gel from the pump spouts and prevents any "stringy" medium from pulling away when the bottle is removed.
To use, simply dispense gel on a room temperature block-holder. Use a cold heat extractor to form a base layer. Dispense additional gel. Place the tissue on top of the gel. Use a cold heat extractor to freeze tissue and form a block.