Learn More
Poltex™ Cryo Glove/Ice Scraper Holder Mount
SDP
Catalog No. 22000783
Change view
Click to view available options
Product Type:
Magnet Mount
Tape Mount
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-000-783 Wall Mount
22-000-780 Magnet Mount
22-000-782 Tape Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-783 Supplier Poltex™ Supplier No. CRYOGLVW
Add to Cart
Add to Cart
Holds cryo gloves and ice scraper. Choose from magnet, tape, or wall mount.
  • Constructed from super durable PETG
  • Choose mounting method: wall mount (screws/anchors included), magnets (powerful magnets indcluded), or VHB Tape (very high bond tape included)
Specifications
Color Transparent
Autoclavable Not Autoclavable
Material PETG
Product Type Wall Mount
Withstand Temperature (Metric) -20°C