Learn More
Andwin Scientific CRYO GLOVES ELBOW SM EA
SDP
Catalog No. 50727523
Encompass_Preferred
The small Elbow Cryo-Gloves are ideal for use in clinical laboratory, bio-medical, blood banks, frozen food processing, and pharmaceutical environments. The gloves can protect users from 135C to 150C. They are water-resistant and are comfortable to wear with its lightweight design.
This item is not returnable. View return policy
Catalog No. 50-727-523 Supplier Andwin Scientific Supplier No. 71076
The small Elbow Cryo-Gloves are ideal for use in clinical laboratory, bio-medical, blood banks, frozen food processing, and pharmaceutical environments. The gloves can protect users from 135C to 150C. They are water-resistant and are comfortable to wear with its lightweight design.