Learn More
Electron Microscopy Sciences Cryo-Gloves Elbow, X-Large, 18" to 20" length
SDP
Catalog No. 50930764
Encompass_Preferred
Cryo-Gloves are essential accessories for those working in ultra cold environments. The gloves provide effective protection in a wide variety of applications from -135 degree C to 150 degree C and provide thermal protection as well. They are water-resistance, comfortable, lightweight and provide excellent dexterity.
Use for ultra low and blast freezers, cold rooms, dry ice handling, liquid nitrogen, liquid oxygen, and liquid helium. Use in clinical laboratories, bio-medical, blood banks, frozen food processing, and pharmaceutical environments.
Size: X-Large
Length: 18" to 20"
This item is not returnable. View return policy
Catalog No. 50-930-764 Supplier Electron Microscopy Sciences Supplier No. 71078
Cryo-Gloves are essential accessories for those working in ultra cold environments. The gloves provide effective protection in a wide variety of applications from -135 degree C to 150 degree C and provide thermal protection as well. They are water-resistance, comfortable, lightweight and provide excellent dexterity.
Use for ultra low and blast freezers, cold rooms, dry ice handling, liquid nitrogen, liquid oxygen, and liquid helium. Use in clinical laboratories, bio-medical, blood banks, frozen food processing, and pharmaceutical environments.
Size: X-Large
Length: 18" to 20"