Learn More
Electron Microscopy Sciences Cryo-Gloves Mid Arm, Large, 14" to 15" length
SDP
Catalog No. 501899459
Encompass_Preferred
Cryo-Gloves are essential accessories for those working in ultra cold environments. The gloves provide effective protection in a wide variety of applications from -135 degree C to 150 degree C and provide thermal protection as well. They are water-resistance, comfortable, lightweight and provide excellent dexterity.
Use for ultra low and blast freezers, cold rooms, dry ice handling, liquid nitrogen, liquid oxygen, and liquid helium. Use in clinical laboratories, bio-medical, blood banks, frozen food processing, and pharmaceutical environments.
Size: Large
Length: 14" to 15"
This item is not returnable. View return policy
Catalog No. 50-189-9459 Supplier Electron Microscopy Sciences Supplier No. 71075
Cryo-Gloves are essential accessories for those working in ultra cold environments. The gloves provide effective protection in a wide variety of applications from -135 degree C to 150 degree C and provide thermal protection as well. They are water-resistance, comfortable, lightweight and provide excellent dexterity.
Use for ultra low and blast freezers, cold rooms, dry ice handling, liquid nitrogen, liquid oxygen, and liquid helium. Use in clinical laboratories, bio-medical, blood banks, frozen food processing, and pharmaceutical environments.
Size: Large
Length: 14" to 15"