Learn More
Sigma Aldrich Fine Chemicals Biosciences Cryo-glovesR size L water-
Catalog No. 502905313
Encompass
Provides warmth flexibility and dexterity when working in extreme environments (-160 to 150C).Style provides additional warmth and comfortResistant to solvents/chemicalsBreathable and static-freeMachine washable/dryable inner liningSuitable for Class 1000 clean room useAsbestos-freeApplications Autoclaves freezers handling liquid nitrogen oxygen or helium from cryogenic atmospheres cold rooms blast freezers filling of cryogenic liquids dry ice handling in research and clinical pharmaceutical laboratories genetic and biotechnology fertility clinics cryogenic grinding and many more in industrial applications.
Catalog No. 50-290-5313 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1835201PAK
Provides warmth flexibility and dexterity when working in extreme environments (-160 to 150C).Style provides additional warmth and comfortResistant to solvents/chemicalsBreathable and static-freeMachine washable/dryable inner liningSuitable for Class 1000 clean room useAsbestos-freeApplications Autoclaves freezers handling liquid nitrogen oxygen or helium from cryogenic atmospheres cold rooms blast freezers filling of cryogenic liquids dry ice handling in research and clinical pharmaceutical laboratories genetic and biotechnology fertility clinics cryogenic grinding and many more in industrial applications.