Learn More
Electron Microscopy Sciences Cryogenic Grid Handling Tool
SDP
Catalog No. 5033956
Encompass_Preferred
This tool has one end which is threaded and fits into the center hole of the Cryogenic Grid Storage Box (where the screw goes in to secure the lid) for moving the box in and out of the cryogenic chamber.
Catalog No. 50-339-56 Supplier Electron Microscopy Sciences Supplier No. 7116550
This tool has one end which is threaded and fits into the center hole of the Cryogenic Grid Storage Box (where the screw goes in to secure the lid) for moving the box in and out of the cryogenic chamber.