Learn More
Mitegen Plastic Cryo Grid Box Gripper Tool For SKU M-CEM-AGG1001, For Secure Transfer of Metal Cryo Grid Boxes with Pin-Type Lids
SDP
Catalog No. NC1816925
Encompass
This Plastic Cryo Grid Box Gripper Tool is specifically designed for the secure and efficient transfer of various grid boxes, particularly the Metal Cryo Grid Box with a pin-type lid (SKU M-CEM-AGG1001). Crafted from durable plastic, this gripper ensures a firm, stable hold on grid boxes, minimizing direct hand contact and reducing the risk of accidental drops or thermal shock to delicate cryogenic samples during transfer between locations. It is ideally suited for moving grid boxes between dewars, workstations, or into automated systems. This tool is intended solely for transferring grid boxes and should not be used to open or unscrew lids, ensuring proper handling procedures are maintained. Its ergonomic design enhances control and safety in cryo-electron microscopy workflows, contributing to streamlined sample management and protecting valuable research materials in ultra-low temperature environments.
Catalog No. NC1816925 Supplier Mitegen Supplier No. MCEMAGT1002
This Plastic Cryo Grid Box Gripper Tool is specifically designed for the secure and efficient transfer of various grid boxes, particularly the Metal Cryo Grid Box with a pin-type lid (SKU M-CEM-AGG1001). Crafted from durable plastic, this gripper ensures a firm, stable hold on grid boxes, minimizing direct hand contact and reducing the risk of accidental drops or thermal shock to delicate cryogenic samples during transfer between locations. It is ideally suited for moving grid boxes between dewars, workstations, or into automated systems. This tool is intended solely for transferring grid boxes and should not be used to open or unscrew lids, ensuring proper handling procedures are maintained. Its ergonomic design enhances control and safety in cryo-electron microscopy workflows, contributing to streamlined sample management and protecting valuable research materials in ultra-low temperature environments.