Learn More
Research Products International Corp CryoFreeze GripRack Cryogenic Storage Racks, 40 Tube Capacity, 5 per Case
SDP
Catalog No. 501537676
Encompass
GripRack Storage Rack is unique, ruggedly designed 40 place rack. Ribs inside the wells of the rack hold stargrip base tubes in position for easy, one-handed opening and closing of tubes. The GripRacks rubber non-slip grips keep the rack secure on the bench or in your hand. High contrast alphanumeric indexing for fast sample tube location and identification.
Catalog No. 50-153-7676 Supplier Research Products International Corp Supplier No. 212607
GripRack Storage Rack is unique, ruggedly designed 40 place rack. Ribs inside the wells of the rack hold stargrip base tubes in position for easy, one-handed opening and closing of tubes. The GripRacks rubber non-slip grips keep the rack secure on the bench or in your hand. High contrast alphanumeric indexing for fast sample tube location and identification.