Learn More
Zen Bio Inc Cryopreserved Human Hepatic Sinusoidal Endothelial Cells, (500,000 cells/vial)
SDP
Catalog No. NC3237687
Encompass
Human Hepatic Non-Parenchymal CellsCryopreserved Human Hepatic Sinusoidal Endothelial Cells, (500,000 cells/vial) Cells have been tested for performance in viability. Cells have tested negative for HIV-1, HIV-2, HbsAG, HbcAB, HCV, and RPR. Cells are additionally tested for CMV.
Catalog No. NC3237687 Supplier Zen Bio Inc Supplier No. HSECF
Human Hepatic Non-Parenchymal CellsCryopreserved Human Hepatic Sinusoidal Endothelial Cells, (500,000 cells/vial) Cells have been tested for performance in viability. Cells have tested negative for HIV-1, HIV-2, HbsAG, HbcAB, HCV, and RPR. Cells are additionally tested for CMV.