Learn More
Cell Applications Inc CRYO HUVEC N 1 AMPOULE
SDP
Catalog No. 50608971
Encompass
Human Umbilical Vein Endothelial Cells (HUVEC) provide an classic model system to study many aspects of endothelial function and disease, such as normal, abnormal and tumor-associated angiogenesis, oxidative stress, hypoxia and inflammation.
Catalog No. 50-608-971 Supplier Cell Applications Inc Supplier No. 20005N
Human Umbilical Vein Endothelial Cells (HUVEC) provide an classic model system to study many aspects of endothelial function and disease, such as normal, abnormal and tumor-associated angiogenesis, oxidative stress, hypoxia and inflammation.