Learn More
GA International Cryogenic Tags for Metal Racks
Supplier:  GA International CID02A2
Encompass_Preferred
Hanging tags for the identification of metal racks and other containers stored long-term in liquid nitrogen Dewars and ultra-low temperature laboratory freezers. Each Cryo-ID-Kit™ includes 12 plastic Cryo-Cards™ with connec¬tion hooks and thermal-transfer cryogenic labels. The supplied NitroTAGTM labels can be printed on using all common thermal-transfer barcode printer brands with alphanumeric text, serialized information, as well as 1D and 2D barcodes. The labels can also be inscribed with permanent markers.
Catalog No. NC1163664