Learn More
Alkali Scientific ASI™ Cryo Label Rolls, 33x13mm for 1.5/2mL Tubes, 1000 per Roll, 1/Roll, Blue
SDP
Catalog No. 503028756
Encompass_Preferred
CRYO LABEL ROLLS 33x13mm for 1.5/2ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 1000, 1 roll/cs
Catalog No. 50-302-8756 Supplier Alkali Scientific Supplier No. LCR12114B
CRYO LABEL ROLLS 33x13mm for 1.5/2ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 1000, 1 roll/cs