Learn More
Alkali Scientific ASI™ Cryo Label Rolls, 38x19mm for General Use, 1000 per Roll, 1/Roll, Blue
SDP
Catalog No. 503027271
Encompass_Preferred
CRYO LABEL ROLLS 38x19mm for General Use We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 1000, 1 roll/cs
Catalog No. 50-302-7271 Supplier Alkali Scientific Supplier No. LCR34112B
CRYO LABEL ROLLS 38x19mm for General Use We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 1000, 1 roll/cs