Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 24x13mm for 0.5mL Tubes White, Assorted, 2975 labels, 20 sheets/Cs
SDP
Catalog No. 503027037
Encompass_Preferred
CRYO LABEL SHEETS 24x13mm for 0.5ml Tubes White We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).  
Catalog No. 50-302-7037 Supplier Alkali Scientific Supplier No. LCL121A
CRYO LABEL SHEETS 24x13mm for 0.5ml Tubes White We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).  