Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 33x13mm for 1.5-2mL Tubes, Green, 1700 labels, 20 sheets/Cs
SDP
Catalog No. 503028834
Encompass_Preferred
CRYO LABEL SHEETS 33x13mm for 1.5-2ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).    
Catalog No. 50-302-8834 Supplier Alkali Scientific Supplier No. LCL12114G
CRYO LABEL SHEETS 33x13mm for 1.5-2ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).    