Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 38x19mm for General Use, White, 1200 labels, 20 sheets/Cs
SDP
Catalog No. 503027582
Encompass_Preferred
CRYO LABEL SHEETS 38x19mm for General Use We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).
Catalog No. 50-302-7582 Supplier Alkali Scientific Supplier No. LCL34112SW
CRYO LABEL SHEETS 38x19mm for General Use We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).