Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 43x19mm for Cryovials, White, 1040 labels, 20 sheets/Cs
SDP
Catalog No. 503028758
Encompass_Preferred
CRYO LABEL SHEETS 43x19mm for Cryovials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 500, 1 Roll/cs
Catalog No. 50-302-8758 Supplier Alkali Scientific Supplier No. LCL34169W
CRYO LABEL SHEETS 43x19mm for Cryovials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 500, 1 Roll/cs