Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 67x25mm for Racks/Boxes, Red, 600 labels, 20 sheets/Cs
SDP
Catalog No. 503027584
Encompass_Preferred
CRYO LABEL SHEETS 67x25mm for Racks/Boxes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).
Catalog No. 50-302-7584 Supplier Alkali Scientific Supplier No. LCL1258R
CRYO LABEL SHEETS 67x25mm for Racks/Boxes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).