Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 9.5mm Dots for 0.5-1.5mL Tubes, Orange, 3840 labels, 20 sheets/Cs
SDP
Catalog No. 503028836
Encompass_Preferred
CRYO LABEL SHEETS 9.5mm Dots for 0.5-1.5ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).  
Catalog No. 50-302-8836 Supplier Alkali Scientific Supplier No. LCL38O
CRYO LABEL SHEETS 9.5mm Dots for 0.5-1.5ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).  