Learn More
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials & Surfaces 1.25" x 0.875" / 31.8mm x 22.2mm Rectangle White From -196°C to +121°C (-321°F to +250°F)
Catalog No. NC2326477
Encompass_Preferred
These cryogenic labels provide an all-in-one solution for cryo storage as they firmly adhere to both ambient and already frozen* surfaces (as low as -80°C/-112°F) without the need for thawing, cure time, or any other special treatment. They maintain excellent long-term adhesion when stored under cryogenic conditions. Due to their special adhesive and conformable design, these cryo labels can be applied onto both flat and curved surfaces, ideal for identifying cryo vials, microtubes, as well as cardboard and polycarbonate storage boxes destined for long-term storage in liquid nitrogen tanks (-196°C/-321°F), ultra-low temperature freezers (-80°C) and dry ice transportation. Perfectly suited for identifying vials destined for immediate immersion in liquid nitrogen as well as labeling already frozen vials without having to thaw them first, thus preserving the biological integrity of the frozen specimens.These permanent cryo labels will also resist elevated temperatures up to +121°C/+249°
Catalog No. NC2326477 Supplier GA International Supplier No. UC9C11WH
These cryogenic labels provide an all-in-one solution for cryo storage as they firmly adhere to both ambient and already frozen* surfaces (as low as -80°C/-112°F) without the need for thawing, cure time, or any other special treatment. They maintain excellent long-term adhesion when stored under cryogenic conditions. Due to their special adhesive and conformable design, these cryo labels can be applied onto both flat and curved surfaces, ideal for identifying cryo vials, microtubes, as well as cardboard and polycarbonate storage boxes destined for long-term storage in liquid nitrogen tanks (-196°C/-321°F), ultra-low temperature freezers (-80°C) and dry ice transportation. Perfectly suited for identifying vials destined for immediate immersion in liquid nitrogen as well as labeling already frozen vials without having to thaw them first, thus preserving the biological integrity of the frozen specimens.These permanent cryo labels will also resist elevated temperatures up to +121°C/+249°