Learn More
Electron Microscopy Sciences Cryo Labels, 2.5cm x 5cm
Supplier: Electron Microscopy Sciences 62047
A special formulated material for use at ultra-low temperatures and in cryo storage conditions, such as liquid nitrogen and oxygen. They are durable and adhere well to plastic as well as cardboard cryogenic storage boxes. They will not peel or shrink. Ten sheets of labels: five preprinted, five plain. Twenty labels/sheet.
Size: 2.5cm x 5cm