Learn More
GA International Cryogenic Lamination Tape 1" x 200' / 25.4mm x 60m Transparent From -196°C to +121°C (-321°F to +250°F)
Catalog No. NC2879380
Encompass_Preferred
Our clear Cryo-Laminate™ tape provides excellent adhesion and can be used to protect existing labels or other printed information on various surfaces, including cardboard, plastic, and metal, from a wide range of temperatures from -196°C up to +121°C. This permanent lamination tape is ideal for identifying containers destined for long-term storage in liquid and vapor phase liquid nitrogen (-196°C/-321°F), ultra-low temperature freezers (-80°C/-112°F), and dry ice transportation. Cryo-Laminate tape can also withstand standard autoclave conditions (+121°C/+249°F, 16psi, 20 minutes) as well as incubation in heated water baths. It protects printed data by safeguarding the existing label from cryo conditions while shielding it from abrasion and scratching.Cryo-Laminate tapes can be applied directly over frozen cryo vials, as well as plastic and cardboard freezer boxes. Moreover, Cryo-Laminate tapes are hand writable using our permanent cryo markers.
Catalog No. NC2879380 Supplier GA International Supplier No. TCA25CP3200
Our clear Cryo-Laminate™ tape provides excellent adhesion and can be used to protect existing labels or other printed information on various surfaces, including cardboard, plastic, and metal, from a wide range of temperatures from -196°C up to +121°C. This permanent lamination tape is ideal for identifying containers destined for long-term storage in liquid and vapor phase liquid nitrogen (-196°C/-321°F), ultra-low temperature freezers (-80°C/-112°F), and dry ice transportation. Cryo-Laminate tape can also withstand standard autoclave conditions (+121°C/+249°F, 16psi, 20 minutes) as well as incubation in heated water baths. It protects printed data by safeguarding the existing label from cryo conditions while shielding it from abrasion and scratching.Cryo-Laminate tapes can be applied directly over frozen cryo vials, as well as plastic and cardboard freezer boxes. Moreover, Cryo-Laminate tapes are hand writable using our permanent cryo markers.