Learn More
Electron Microscopy Sciences Cryo-Lok Cyrogenic Vial, 2.5 ml, Non-Sterile
SDP
Catalog No. 50949574
Encompass_Preferred
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures. The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal. Graduated in 0.5 ml increments. For use in vapor phase of liquid nitrogen.
Volume: 2.5 ml
Sterile: Non-Sterile
Catalog No. 50-949-574 Supplier Electron Microscopy Sciences Supplier No. 72652
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures. The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal. Graduated in 0.5 ml increments. For use in vapor phase of liquid nitrogen.
Volume: 2.5 ml
Sterile: Non-Sterile