Learn More
Electron Microscopy Sciences Cryo Markers 6 per pack/case
SDP
Catalog No. 501899074
Encompass_Preferred
For permanently marking cloth, cardboard, and plastic. It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).
Catalog No. 50-189-9074 Supplier Electron Microscopy Sciences Supplier No. 62051
For permanently marking cloth, cardboard, and plastic. It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).