Learn More
Med Vet International Jorvet Cryogun Attachments, 1mm Mini Closed Probe, J0810GA
Catalog No. 5000495808
Encompass_Preferred
Used to treat eyelid margin and corneal lesions. Gently curved to allow visualization and precise control.
Catalog No. 50-004-95808 Supplier Med Vet International Supplier No. JOR810GA
Used to treat eyelid margin and corneal lesions. Gently curved to allow visualization and precise control.