Learn More
Med Vet International Jorvet Cryogun Attachments, 6mm Mini Closed Probe, J0810GF
Catalog No. 5000495835
Encompass_Preferred
Used to treat eyelid margin and corneal lesions. Gently curved to allow visualization and precise control.
Catalog No. 50-004-95835 Supplier Med Vet International Supplier No. JOR810GF
Used to treat eyelid margin and corneal lesions. Gently curved to allow visualization and precise control.