Learn More
IMEB, Inc Tissue-Tek Cryomolds, 5X15X5mm. 100 per Pack
SDP
Catalog No. NC0680250
Encompass
Tissue-Tek Cryomold create a uniformed shaped, flat-surface specimen block with O.C.T. Compound for frozen sectioning. Disposable plastic molds fit Cryobar freezing wells. Available in 3 sizes: standard, fits directly into the Cryobar freezing wells, biopsy and standard square require adapters for optimum uniform freezing in the Cryobar. 100 per pack
Catalog No. NC0680250 Supplier IMEB, Inc Supplier No. 4730
Tissue-Tek Cryomold create a uniformed shaped, flat-surface specimen block with O.C.T. Compound for frozen sectioning. Disposable plastic molds fit Cryobar freezing wells. Available in 3 sizes: standard, fits directly into the Cryobar freezing wells, biopsy and standard square require adapters for optimum uniform freezing in the Cryobar. 100 per pack