Learn More
Andwin Scientific Cryo Mold Intermediate 15x15x5 mm 100/pack
SDP
Catalog No. NC9642669
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 5 mm. It is sold in a pack of 100.
Catalog No. NC9642669 Supplier Andwin Scientific Supplier No. 6253415
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 15 mm x 15 mm x 5 mm. It is sold in a pack of 100.