Learn More
Research Products International Corp Cryo Marker for Freezing Purple 3/pk
SDP
Catalog No. 502252989
Encompass
Cryo Marker is ideal for freezing applications. Big tip is perfect for clearly marking various surfaces. Dries quickly in 5 - 7 minutes for permanent, fade resistant, waterproof marking.  Use directly on labels, container at room temperature before storing in liquid nitrogen (-196°C) or deep freezing (-80°C) applications.
Alcohol and water resistant
Thermal resistance -196°C to +200°C
Big tip
This item is not returnable. View return policy
Catalog No. 50-225-2989 Supplier Research Products International Corp Supplier No. 248512PU
Cryo Marker is ideal for freezing applications. Big tip is perfect for clearly marking various surfaces. Dries quickly in 5 - 7 minutes for permanent, fade resistant, waterproof marking.  Use directly on labels, container at room temperature before storing in liquid nitrogen (-196°C) or deep freezing (-80°C) applications.
Alcohol and water resistant
Thermal resistance -196°C to +200°C
Big tip