Learn More
GA International Writable Cryogenic Multi-Color Dots 0.35" / 9mm Circle 8 assorted colors From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC1561135
Encompass_Preferred
These small-diameter, waterproof round cryo labels are designed for microtube tops, enabling clear identification of tubes stored in low-temperature environments. Writable and compatible with cryogenic markers, they feature a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen*. Supplied in a multi-color roll within a convenient dispenser box for smooth dispensing and a clean, organized lab bench. They are ideal for easy color-coding and quick sample identification.* Use in liquid nitrogen is subject to prior verification of compatibility between the labels and the intended vial cap material.
Catalog No. NC1561135 Supplier GA International Supplier No. LTR9X8A
These small-diameter, waterproof round cryo labels are designed for microtube tops, enabling clear identification of tubes stored in low-temperature environments. Writable and compatible with cryogenic markers, they feature a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen*. Supplied in a multi-color roll within a convenient dispenser box for smooth dispensing and a clean, organized lab bench. They are ideal for easy color-coding and quick sample identification.* Use in liquid nitrogen is subject to prior verification of compatibility between the labels and the intended vial cap material.