Learn More
Cole-Parmer Nalgene 6313-0010 Cryoware markers, 4/pack (One each of four colors (red, blue, green, and black).
Catalog No. NC9919507 Shop All Cole-Parmer Products
Encompass_Preferred
Mark vials, and cardboard and plastic cryogenic boxes with these Cryoware markers. They will not smudge or fade at temperatures as low as -196C, and mark on surfaces free of condensation. The extra fine tip markers allow writing on small surfaces, such as cryogenic vials and tubes.
Catalog No. NC9919507 Supplier Cole-Parmer Supplier No. 0996440
Mark vials, and cardboard and plastic cryogenic boxes with these Cryoware markers. They will not smudge or fade at temperatures as low as -196C, and mark on surfaces free of condensation. The extra fine tip markers allow writing on small surfaces, such as cryogenic vials and tubes.