Learn More
Electron Microscopy Sciences Cryo Ultramicrotome Sample Pin, 2 x 10 mm
SDP
Catalog No. 501929106
Encompass
This cryo pin has a diameter of 2 mm and is compatible with the Leica and RMC cryo ultramicrotomes. Made from cryo compatible material. Concentric grooves on the sample surface ensure better adhesion. Reversed tapered head with a diameter of 3.5 mm and 10 mm long.
Size: 2 x 10 mm
Catalog No. 50-192-9106 Supplier Electron Microscopy Sciences Supplier No. 7595906
This cryo pin has a diameter of 2 mm and is compatible with the Leica and RMC cryo ultramicrotomes. Made from cryo compatible material. Concentric grooves on the sample surface ensure better adhesion. Reversed tapered head with a diameter of 3.5 mm and 10 mm long.
Size: 2 x 10 mm