Learn More
Research Products International Corp Cryo-LazrTag Waterproof Cryogenic Labels for Laser Printers, 0.94 x 0.5 Inch Label, Mint, 2688 Labels per Pack
SDP
Supplier:  Research Products International Corp 247182M
Encompass
Pre-cut, peel-off labels fit cryogenic tubes/vials and are ideal for long-term storage in liquid nitrogen and freezers. Waterproof labels are scuff-resistant, smear-resistant and adhere to all plastics and withstand multiple freeze-thaw cycles. These labels are waterproof and can withstand multiple freeze-thaw cycles without detaching or delaminating, scuff-resistant and smear-resistant. Easy to print with all major brands of laser printers, laser copiers and write on with permanent markers. The ergonomic layout allows the maximum use of label space.
Catalog No. 50-153-7698