Learn More
Shields Dina Corporation CRYO PROTECTION GLOVE BLUE L
Catalog No. NC3243301
Encompass
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size Large
Catalog No. NC3243301 Supplier Shields Dina Corporation Supplier No. CRYOLNGL
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size Large