Learn More
Shields Dina Corporation CRYO PROTECTION GLOVE BLUE M
Catalog No. NC3243290
Encompass
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size Medium
Catalog No. NC3243290 Supplier Shields Dina Corporation Supplier No. CRYOLNGM
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size Medium