Learn More
Shields Dina Corporation CRYO PROTECTION GLOVE BLUE XL
Catalog No. NC3243304
Encompass
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size X-Large
Catalog No. NC3243304 Supplier Shields Dina Corporation Supplier No. CRYOLNGXL
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size X-Large