Learn More
Shields Dina Corporation CRYO PROTECTION GLOVE BLUE XXL
Catalog No. NC3243306
Encompass
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size XX-Large
Catalog No. NC3243306 Supplier Shields Dina Corporation Supplier No. CRYOLNGXXL
TempShield, Cryo Protection Glove, Blue, Elbow Length, WaterproofLiner, Cryogenic Protection for Ultra-Cold Applications Down to -196 C/-320 F, Size XX-Large