Learn More
Electron Microscopy Sciences CRYO-PRTCTN GRP SFTY KT S 42IN
SDP
Catalog No. 502413039
Encompass_Preferred
Cryo-Protection Grip Safety Kit is an essential PPE for those working with cryogenic liquids. Where there is a chance of exposure to splashes of lquids these kits are useful in dispersing or transferring cryogenic liquids, removing samples from cryogenic liquids, operation involving cryogenic continers. It provides cryogenic protection for ultra cold application down to -196 deg celcius. With a multi layer construction the material provides maximum thermal protection, flexibilty, and dextrity while maintaining both proper function and safety. The gloves has a grip palm for improved dexterity handling of small objects and a thermal inner lining that removes moisture away from hands and provides comfort over extended periods. Safety certfied to meed EU standards for CE CAT. III: EN 511, EN 388 and EN 420. Kit not intended for immersion in liquid nitrogen or other cryogenic liquids.
Catalog No. 50-241-3039 Supplier Electron Microscopy Sciences Supplier No. 7108309
Cryo-Protection Grip Safety Kit is an essential PPE for those working with cryogenic liquids. Where there is a chance of exposure to splashes of lquids these kits are useful in dispersing or transferring cryogenic liquids, removing samples from cryogenic liquids, operation involving cryogenic continers. It provides cryogenic protection for ultra cold application down to -196 deg celcius. With a multi layer construction the material provides maximum thermal protection, flexibilty, and dextrity while maintaining both proper function and safety. The gloves has a grip palm for improved dexterity handling of small objects and a thermal inner lining that removes moisture away from hands and provides comfort over extended periods. Safety certfied to meed EU standards for CE CAT. III: EN 511, EN 388 and EN 420. Kit not intended for immersion in liquid nitrogen or other cryogenic liquids.