Learn More
Greiner Bio-One Cryo.s Cryogenic Storage Tube with Datamatrix Code
GREENER_CHOICE
Catalog No. 07000275 Shop All Greiner Bio-One Products
Change view
Click to view available options
Capacity (Metric):
2 mL
Catalog No. Capacity (Metric)
07-000-275 2 mL
1 options
Catalog No. 07-000-275 Supplier Greiner Bio-One Supplier No. 1222632DG
Add to Cart
Add to Cart
Laser-written ECC200 Datamatrix Coding Service for cryogenic storage tubes.
  • Strong resistance to mechanical and chemical damage
  • High data capacity
  • Sequential sample IDs or consistent characters
  • Reed-Solomon error correction for high redundancy of encrypted information
TRUSTED_SUSTAINABILITY
Specifications
Description Datamatrix Code: 14 x 14, Sequence: G200000001, G200000002
Closure Color Natural
Autoclavable Not Autoclavable
Capacity (Metric) 2 mL
Bottom Shape Round (Starfoot)
Material Polypropylene
Thread Style Internal
Includes Writing area
Sterility Sterile
Graduated Yes
Closure Type Screw Cap
Product Type Cryogenic Storage Tube