Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Cryo.s Round Bottom 4 mL Polypropylene Tube with External Thread Cap
GREENER_CHOICE

Catalog No. 07000437 Shop All Greiner Bio-One Products
Change view
Click to view available options
Closure Color:
Blue
Green
Natural
Red
Yellow
Includes:
150 Inserts
DataMatrix ECC 200, 150 Inserts
ID Writing Area, 150 Inserts
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Includes
07-000-437 Natural 150 Inserts
07-000-438 Natural ID Writing Area, 150 Inserts
07-000-439 Yellow ID Writing Area, 150 Inserts
07-000-440 Red ID Writing Area, 150 Inserts
07-000-260 Blue ID Writing Area, 150 Inserts
07-000-261 Green ID Writing Area, 150 Inserts
07-000-276 Natural DataMatrix ECC 200, 150 Inserts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 07-000-437 Supplier Greiner Bio-One Supplier No. 127261
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.

  • With or without ID writing area
  • With Star foot base
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 100 white cap inserts
  • Polyethylener case
  • Dia. x Height: 12.4 x 83mm

Service Plan

No

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Natural
Capacity (Metric) 4 mL
Bottom Shape Round (Starfoot)
Material Polypropylene
Thread Style External
Includes 150 Inserts
Length (Metric) 83 mm
Packaging Polyethylene case
Sterility Sterile
Closure Type Threaded Screw Cap
Diameter (Metric) 12.4 mm
Product Type Cryogenic Storage Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.