Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Cryo-Safe™ Cold Box
SCI_ED

Catalog No. s90150 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S90150 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S90150 Supplier Bel-Art™ Supplier No. F188470002/EMD
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Made of molded polystyrene. Bel Art™ Cryo-Safe™ Cold Box. Has an internal 12-place support for 1.5 mL microcentrifuge tubes..

  • When container is taken out of a -20°C (-4°F) freezer, it maintains a -15°C (-5°F) temperature for 1.5 hours, and will remain below 0°C (32°F) for four hours.
  • Consists of a molded polystyrene container with cover
  • Internal 12-place support for 0.05 oz. (1.5 mL) microcentrifuge tubes
  • Space surrounding tubes is filled with a sealed-in material to maintain the low temperature required

Specifications

Capacity (English) 0.05 oz.
Includes Cover
For Use With (Application) Safeguards enzymes and thermolabile substances from thermal degradation
Material Polystyrene
Type Cryo-Safe Cold Box
Color White
Capacity (Metric) 1.5 mL
Holds 12 Microcentrifuge Tubes (15 mL)
Autoclavable Not Autoclavable
Temperature Control No
Temperature Range -15°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.