Learn More
Research Products International Corp Cryo-Safe Cooler with Clear Lid, Holds 0.5 or 1.5ml Tubes, 12 Tube Capacity
SDP
Catalog No. 50999132
Encompass
Bench top sized cooler holds twelve 0.5 or 1.5ml microcentrifuge tubes at a temperature below 0ºC for approximately 2 1/2 hours. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Use included insert to hold 0.5 ml microcentrifuge tubes. Store at -20ºC for a few hours until the temperature of the cooler falls below 0ºC. Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe. A clear polycarbonate lid shows positions of tubes; keyed to correspond with positions printed on cooler.
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-999-132 Supplier Research Products International Corp Supplier No. 142414
Bench top sized cooler holds twelve 0.5 or 1.5ml microcentrifuge tubes at a temperature below 0ºC for approximately 2 1/2 hours. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Use included insert to hold 0.5 ml microcentrifuge tubes. Store at -20ºC for a few hours until the temperature of the cooler falls below 0ºC. Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe. A clear polycarbonate lid shows positions of tubes; keyed to correspond with positions printed on cooler.
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.