Learn More
Research Products International Corp Cryo-Safe Cooler w/ Gel Filled Lid, 32 Tube Capacity
SDP
Catalog No. 501977340
Encompass
Benchtop sized cooler holds thirty-two 0.5 or 1.5 ml microcentrifuge tubes at a temperature below 0ºC for approximately 3 hours. Use included insert to hold 0.5 ml microcentrifuge tubes. Store at -20ºC for a few hours until the temperature of the cooler falls below 0ºC. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe. .
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7340 Supplier Research Products International Corp Supplier No. 142416
Add to Cart
Add to Cart
Benchtop sized cooler holds thirty-two 0.5 or 1.5 ml microcentrifuge tubes at a temperature below 0ºC for approximately 3 hours. Use included insert to hold 0.5 ml microcentrifuge tubes. Store at -20ºC for a few hours until the temperature of the cooler falls below 0ºC. Ideal for protecting valuable samples stored in a freezer with defrost cycle. Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe. .
WARNING  This product can expose you to chemicals including Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.