Learn More
Sigma Aldrich Fine Chemicals Biosciences Cryo-SafeTM vial storage b
Catalog No. 502900196
Encompass
The Cryo-Safe vial storage box can be used to transfer or ship vial or tubes. The storage box is rugged polycarbonate boxes that can withstand temperatures of -196 C to 121 C.Select box to hold 1.2 2.0 or 5.0 mL cryogenic vialsTransparent lids are keyed for alignment and include numbering grid for sample identityDimension 13.3 x 13.3 x 9.5cm (5A¼ x 5A¼ x 3A¾)
Catalog No. 50-290-0196 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7567844EA
The Cryo-Safe vial storage box can be used to transfer or ship vial or tubes. The storage box is rugged polycarbonate boxes that can withstand temperatures of -196 C to 121 C.Select box to hold 1.2 2.0 or 5.0 mL cryogenic vialsTransparent lids are keyed for alignment and include numbering grid for sample identityDimension 13.3 x 13.3 x 9.5cm (5A¼ x 5A¼ x 3A¾)