Learn More
Taylor Wharton Cryo-Sentry Level Alarm for HC20 & HC34 Liquid Nitrogen Systems
Catalog No. NC0701914
Encompass_Preferred
Low Level Alarm to protect valuable biological samples. This alarm will sound promptly to alert you when more liquid nitrogen is needed at approximatley 1/4 volume.
Catalog No. NC0701914 Supplier Taylor Wharton Supplier No. R0348C15
Low Level Alarm to protect valuable biological samples. This alarm will sound promptly to alert you when more liquid nitrogen is needed at approximatley 1/4 volume.