Learn More
Electron Microscopy Sciences Cryo-Specimen Pins; Bullseye
SDP
Catalog No. 50930303
Encompass
Aluminum specimen mount pin for mounting samples for ultramicrotomy.
Catalog No. 50-930-303 Supplier Electron Microscopy Sciences Supplier No. 70446
Aluminum specimen mount pin for mounting samples for ultramicrotomy.