Learn More
Research Products International Corp Cryogenic Storage Box, Hinged Lid, 100 Tube Capacity, Natural Color, 5 per Case
SDP
Catalog No. 50550189
Encompass
Durable polypropylene storage boxes withstand repeated low temperature storage. Plastic boxes will not deterioate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Accommodates all hinged and screw cap 1.5-2.0ml micro-tubes.
Catalog No. 50-550-189 Supplier Research Products International Corp Supplier No. 145635
Durable polypropylene storage boxes withstand repeated low temperature storage. Plastic boxes will not deterioate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Accommodates all hinged and screw cap 1.5-2.0ml micro-tubes.