Learn More
Research Products International Corp Cryogenic Storage Rack, Hinged Lid, 50 Tube Capacity, Assorted Colors, 5 per Case
SDP
Catalog No. 50550188
Encompass
Durable polypropylene storage boxes withstand repeated low temperature storage. Will not deterioate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Available in assorted colors (green, red, blue, orange, yellow). Accommodates all hinged and screw cap 1.5-2.0ml micro-tubes.
Catalog No. 50-550-188 Supplier Research Products International Corp Supplier No. 145632
Durable polypropylene storage boxes withstand repeated low temperature storage. Will not deterioate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Available in assorted colors (green, red, blue, orange, yellow). Accommodates all hinged and screw cap 1.5-2.0ml micro-tubes.