Learn More
New England Biolabs, Inc. Cryogenic Cold Storage and Freezer Boxes – 4 boxes
SDP
Catalog No. 502581803
Encompass
The Cryogenic Cold Storage Box from New England Biolabs is a cryo freezer box designed to protect and store microcentrifuge vials and tubes at -80°C to room temperature (not recommended for liquid nitrogen storage). These cryo boxes are composed of fiberboard/cardboard material and can hold up to 81 items with the included adjustable slats that can be moved or removed to adapt to different tube sizes. The boxes feature a labeled grid with numbers and letters on the sides and the bottom for easy reference and organization. The unique hinged lid is fastened to the box to minimize the risk of separation. The design features colorful butterfly graphics with space to personalize and label the outside of the box for visibility and tracking.
Catalog No. 50-258-1803 Supplier New England Biolabs, Inc. Supplier No. T1041S
The Cryogenic Cold Storage Box from New England Biolabs is a cryo freezer box designed to protect and store microcentrifuge vials and tubes at -80°C to room temperature (not recommended for liquid nitrogen storage). These cryo boxes are composed of fiberboard/cardboard material and can hold up to 81 items with the included adjustable slats that can be moved or removed to adapt to different tube sizes. The boxes feature a labeled grid with numbers and letters on the sides and the bottom for easy reference and organization. The unique hinged lid is fastened to the box to minimize the risk of separation. The design features colorful butterfly graphics with space to personalize and label the outside of the box for visibility and tracking.